missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pinocembrin, MedChemExpress
Description
Pinocembrin ((+)-Pinocoembrin) is a flavonoid found in propolis, acts as a competitive inhibitor of histidine decarboxylase, and is an effective anti-allergic agent, with antioxidant, antimicrobial and anti-inflammatory properties.
Specifications
Specifications
| Chemical Name or Material | Pinocembrin |
| CAS | 480-39-7 |
| Color | Light Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C15H12O4 |
| Synonym | (+)-Pinocoembrin Dihydrochrysin Galangin flavanone |
| Solubility Information | DMSO : 83.33 mg/mL (325.19 mM; Need ultrasonic) |
| SMILES | O=C1C[C@@H](C2=CC=CC=C2)OC3=CC(O)=CC(O)=C13 |
| Molecular Weight (g/mol) | 256.25 |
| Quantity | 25 mg |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?