missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Pinin Polyclonal antibody specifically detects Pinin in Human,Rat samples. It is validated for ELISA,Western Blot
Specifications
Specifications
| Antigen | Pinin |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500 - 1:2000, ELISA |
| Formulation | PBS (pH 7.3), 50% glycerol |
| Gene Alias | 140 kDa nuclear and cell adhesion-related phosphoprotein, Desmosome-associated protein, Domain-rich serine protein, DRS protein, DRSP, DRSSR-like protein, Melanoma metastasis clone A protein, MEMA, neutrophil protein, Nuclear protein SDK3, pinin, pinin, desmosome associated protein, SDK3 |
| Host Species | Rabbit |
| Immunogen | A synthetic peptide corresponding to a sequence within amino acids 1-100 of human Pinin (NP_002678.2).,, Sequence:, MAVAVRTLQEQLEKAKESLKNVDENIRKLTGRDPNDVRPIQARLLALSGPGGGRGRGSLLLRRGFSDSGGGPPAKQRDLEGAVSRLGGERRTRRESRQES |
| Purification Method | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?