Learn More
Pimelic Diphenylamide 106, MedChemExpress
Product Code. 16453717
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16453717 50 mg 50mg
16440208 5 mg 5mg
16427606 100 mg 100mg
16417748 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 16453717 Supplier Medchem Express Supplier No. HY19348
MedChemExpress Pimelic Diphenylamide 106 is a slow, tight-binding inhibitor of class I HDAC (HDAC 1, 2, and 3, with IC50 values of 150 nM , 760nM, and 370 nM, respectively), demonstrating no activity against class II HDACs.
Pimelic Diphenylamide 106 is a slow, tight-binding inhibitor of class I HDAC (HDAC 1, 2, and 3, with IC50 values of 150 nM , 760nM, and 370 nM, respectively), demonstrating no activity against class II HDACs.
Specifications
Chemical Name or Material Pimelic Diphenylamide 106
Physical Form Solid
CAS 937039-45-7
Color White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C20H25N3O2
For Use With (Application) Cancer-programmed cell death
Synonym RGFA-8 TC-H 106 Histone Deacetylase Inhibitor VII
Solubility Information DMSO : 125 mg/mL (368.26 mM; ultrasonic and warming and heat to 60°C)
SMILES O=C(NC1=CC=CC=C1N)CCCCCC(NC2=CC=C(C)C=C2)=O
Molecular Weight (g/mol) 339.43
Formula Weight 339.43
Percent Purity 98.0%
Grade Research