missing translation for 'onlineSavingsMsg'
Learn More

PIGZ Antibody [DyLight 405], Novus Biologicals™

Product Code. 30764553 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30764553 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30764553 Supplier Novus Biologicals Supplier No. NBP347144V

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PIGZ Polyclonal antibody specifically detects PIGZ in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen PIGZ
Applications ELISA, Immunohistochemistry, Western Blot, Immunocytochemistry/Immunofluorescence
Classification Polyclonal
Conjugate DyLight 405
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias class Z, EC 2.4.1, EC 2.4.1.-, FLJ12768, GPI mannosyltransferase 4, GPI mannosyltransferase IV, GPI-MT-IV, hSMP3, phosphatidylinositol glycan anchor biosynthesis, class Z, Phosphatidylinositol-glycan biosynthesis class Z protein, PIG-Z, SMP3 mannosyltransferase, SMP3SMP3 homolog
Host Species Rabbit
Immunogen phosphatidylinositol glycan anchor biosynthesis, class Z. (Uniprot# Q86VD9)
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Endocrinology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 80235
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.