missing translation for 'onlineSavingsMsg'
Learn More

Pigment Yellow 83 (Technical Grade), TRC

Product Code. 30179641
Change view
Click to view available options
Quantity:
1 g
50 g
100 g
Unit Size:
100g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30179641 50 g 50g
30179338 100 g 100g
30179208 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30179641 Supplier Toronto Research Chemicals Supplier No. TRCP43781550G
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5567-15-7
Molecular Formula C36H32Cl4N6O8
Molecular Weight (g/mol) 818.49
Synonym 2,2'-[(3,3'-dichloro[1,1'-biphenyl]-4,4'-diyl)bis(azo)]bis[N-(4-chloro-2,5-dimethoxyphenyl)-3-oxo-butanamide;2,2'-[(3,3'-Dichloro[1,1'-biphenyl]-4,4'-diyl)bis(2,1-diazenediyl)]bis[N-(4-chloro-2,5-dimethoxyphenyl)-3-oxobutanamide];2,2'-[(3,3'-Dichloro[1,1'-biphenyl]-4,4'-diyl)bis(azo)]bis[N-(4-chloro-2,5-dimethoxyphenyl)-3-oxobutanamide, Aquafine Yellow E 12, C.I. 21108, C.I. Pigment Yellow 83;Chromofine Yellow 2700(L), Colanyl Yellow HRD, Disazo Yellow, FG 1842, Graphtol Yellow RCL, HR 70 Yellow, Helio Fast Yellow HRN, Hostafine Yellow HR;Hydrocolor Yellow, Imperial yellow, Imperon Gold Yellow K-RN, Imperon Golden Yellow K-RN, Irgalite Yellow B 3R, Irgalite Yellow B 3RN, Lionol Yellow 1805G;Lionol Yellow 1823G, Lionol Yellow FG 1842, Lionol Yellow NBR, Microlen Yellow B 3RS-MC, Microlith Yellow 2R-A, Microlith Yellow 2R-K, Microlith Yellow 2R-WA;Novoperm Yellow HR, Novoperm Yellow HR 70, Novoperm yellow HR 02, PV Fast Yellow HR, PV Yellow HR, PY 83, Pergantine Yellow 2RN-LA, Permanent Yellow HR;Permanent Yellow HR 11-1400, Permanent Yellow HR 70, Permanent Yellow HR Extra, Pigment Yellow 83, Sanyo Pigment Yellow IRC, Sanyo Pigment Yellow TRG;Segnale Light Yellow R, Seikafast Yellow 2700, Seikafast Yellow 2700B, Seikafast Yellow 2720, Seikafast Yellow 2770, Special Yellow HR, Spectra Pac Yellow 83;Sumitone Fast Yellow HRT, Sunbrite Yellow 83, Sunbrite Yellow 83 275-0570;Sunfast Yellow 83, Symuler Fast Yellow 4173, Symuler Fast Yellow 4181, Symuler Fast Yellow 4181NR, Symuler Fast Yellow HFS, Vulcan Fast Yellow HRD
IUPAC Name 2-[(E)-[2-chloro-4-[3-chloro-4-[(E)-[1-(4-chloro-2,5-dimethoxyanilino)-1,3-dioxobutan-2-yl]diazenyl]phenyl]phenyl]diazenyl]-N-(4-chloro-2,5-dimethoxyphenyl)-3-oxobutanamide
SMILES COc1cc(NC(=O)C(N=Nc2ccc(cc2Cl)c3ccc(N=NC(C(=O)C)C(=O)Nc4cc(OC)c(Cl)cc4OC)c(Cl)c3)C(=O)C)c(OC)cc1Cl

Specifications

Quantity 50 g
InChI Formula InChI=1S/C36H32Cl4N6O8/c1-17(47)33(35(49)41-27-15-29(51-3)23(39)13-31(27)53-5)45-43-25-9-7-19(11-21(25)37)20-8-10-26(22(38)12-20)44-46-34(18(2)48)36(50)42-28-16-30(52-4)24(40)14-32(28)54-6/h7-16,33-34H,1-6H3,(H,41,49)(H,42,50)
Formula Weight 816.1036
Chemical Name or Material Pigment Yellow 83 (Technical Grade)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.