missing translation for 'onlineSavingsMsg'
Learn More

PIGB Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

145.00 EUR - 410.00 EUR

Specifications

Antigen PIGB
Concentration 0.25 mg/mL
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16810424
View Documents
Proteintech
12612-1-AP-20UL
20 μL
145.00 EUR
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16800424
View Documents
Proteintech
12612-1-AP-150UL
150 μL
410.00 EUR
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Mannosyltransferase involved in glycosylphosphatidylinositol-anchor biosynthesis. Transfers the third alpha-1, 2-mannose to Man2-GlcN-acyl-PI during GPI precursor assembly.
Specifications

Specifications

PIGB
Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Rat, Human, Mouse
Q92521, Q9JJQ0
315807, 55981, 9488
PIGB Fusion Protein Ag3244
Primary
-20°C
PIGB
0.25 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
GPI mannosyltransferase 3, GPI mannosyltransferase III, GPI MT III, PIG B, PIGB
Pigb
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.