missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PIGB Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12612-1-AP-150UL
This item is not returnable.
View return policy
Description
Mannosyltransferase involved in glycosylphosphatidylinositol-anchor biosynthesis. Transfers the third alpha-1, 2-mannose to Man2-GlcN-acyl-PI during GPI precursor assembly.Specifications
| PIGB | |
| Polyclonal | |
| Unconjugated | |
| PIGB | |
| GPI mannosyltransferase 3, GPI mannosyltransferase III, GPI MT III, PIG B, PIGB | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 315807, 55981, 9488 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.25 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q92521, Q9JJQ0 | |
| Pigb | |
| PIGB Fusion Protein Ag3244 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction