Learn More
VELP Scientifica Pierceable Septums
Product Code. 30003203 Shop All Velp Products
Change view
Click to view available options
Type:
Pierceable Septums
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
30003203 Pierceable Septums Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30003203 Supplier VELP Scientifica Supplier No. A00000487
  • ONLINE EXCLUSIVE PRICE
    68.44 EUR
    62.00 EUR / Pack of 10
    Save 6.44 EUR
    9% Off
Estimated Shipment: 04-11-2026
Pierceable septums. 10 pcs.
  • Pierceable septums compatible with the VELP CN 802 Carbon/Nitrogen elemental Analyzer liquid kit
Specifications
Type Pierceable Septums