missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sulfo-SBED Biotin Label Transfer Reagent, No-Weigh™ Format

Product Code. 11899240 Shop All Thermo Scientific Products
Click to view available options
Quantity:
10 mg
8 x 1 mg
10 x 1 mg
Unit Size:
10mg
Pack of 10
Pack of 8
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Product Code. 11899240

Brand: Thermo Scientific™ 33033

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Chemically tag purified proteins at primary amines to covalently crosslink and transfer biotinylation to protein interaction binding partners.

Thermo Scientific™ Pierce Sulfo-SBED Biotin Label Transfer Reagent is a multifunctional reagent for labeling a purified protein and then covalently transferring the attached biotin tag onto specific interactors of that protein.

Ready-to-use kit eliminates the need to prepare the necessary buffers or purchase the critical reagents necessary to perform a label transfer experiment. Key reagents, Sulfo-SBED and DTT, are supplied in unique, no-weigh, single-use packaging.

Highlights:

  • Sulfo-NHS ester reacts with primary amines at pH 7-9
  • Aryl azide conjugates randomly when activated with UV light
  • Cleavable disulfide bond allows biotin label transfer
  • Discover protein interactions without radiolabeling

Includes:

No-Weigh Sulfo-SBED Label Transfer Reagent, 8 × 1mg; BupH Phosphate Buffered Saline, 1 Pk. (makes 500mL); Label Transfer Buffer (20X), 200mL (makes 4L); Streptavidin-HRP Conjugate, 0.1mg; No-Weigh Dithiothreitol (DTT), 8 × 7.7mg; Slide-A-Lyzer MINI Dialysis Units plus Float, 10K MWCO, 10-100μL, 10 units/Pk.

TRUSTED_SUSTAINABILITY

Specifications

Format Powder
Quantity 10 mg
Product Type Crosslinker
Solubility Water
Content And Storage Upon receipt store at 4°C.
Labeling Method Chemical Labeling, Label Transfer
Reactive Moiety Sulfo-NHS Ester, Aryl Azide
Product Line Pierce

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.