Learn More
Thermo Scientific™ Pierce™ SATP (N-succinimidyl-S-acetylthiopropionate)
Product Code. 10320974 Shop All Thermo Scientific Products
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Product Code. 10320974
Brand: Thermo Scientific™ 26100
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Short (4.1A), amine-reactive (NHS ester) sulfhydryl-addition reagent; creates stable (protected yet exposable) -SH group for protein crosslinking strategies.
Thermo Scientific™ Pierce SATP is a variant (4.1 Å spacer arm) of SATA, a sulfhydryl-addition reagent for covalent modification of primary amines to facilitate controlled, heterobifunctional crosslinking.
Highlights:
  • Reacts with primary amines to add protected sulfhydryls
  • An easy deprotection step generates a free sulfhydryl group
TRUSTED_SUSTAINABILITY
Specifications
Chemical Reactivity Amine
Content And Storage Upon receipt store desiccated at -20°C. Product is shipped at ambient temperature.
Label or Dye Sulfhydryl Group
Label Type Conjugating Groups
Product Line Pierce
Crosslinker Type Heterobifunctional
Reactive Moiety Active Ester, Succinimidyl Ester, NHS Ester
Spacer Medium (10 to 30 Å)
Form Powder
Reagent Type SATP
Quantity 50 mg
Solubility DMF, DMSO
Product Type Crosslinker