Learn More
Thermo Scientific™ MALDI Matrix Sample Pack, Single-Use
Product Code. 11865834 Shop All Thermo Scientific Products
Click to view available options
Quantity:
24 tubes
Unit Size:
Pack of 8
Product Code. 11865834
Brand: Thermo Scientific™ 90035
Highly purified, recrystallized CHCA, DHB and SA reagents in single-use tubes for mass spectrometry by matrix-assisted laser desorption ionization (MALDI).
Thermo Scientific™ MALDI Matrices are highly purified, recrystallized reagents packaged in convenient single-use tubes sufficient for analyzing up to 96 samples by matrix assisted laser desorption ionization.
The MALDI method for MS peptide analysis is now easier to perform with single-use, highly pure MALDI matrices that are exceptionally convenient to use. Matrix assisted laser desorption ionization (MALDI) matrices produce the cleanest mass spectrometric (MS) spectra when they are recrystallized and prepared daily, which is inconvenient and typically results in significant waste. The packaging of purified, recrystallized alpha-cyano-4-hydroxy-cinnamic acid (CHCA), sinapinic acid (SA) and 2,5-dihydroxybenzoic acid (DHB) MALDI matrices makes preparation of high-quality MALDI reagents easy.
Highlights:
  • Highly purified, recrystallized reagent
  • Packaged in convenient single-use tubes sufficient for analyzing up to 96 samples to save time and reduce waste
  • Provides accurate quantity of matrix every time
TRUSTED_SUSTAINABILITY
Specifications
Description Pierce MALDI Matrix Sample Pack
For Use With (Equipment) Mass Spectrometer
Product Type MALDI Matrix
Content And Storage Upon receipt store product at 4°C.
Starting Material Small Molecules
Final Product Type Small Molecules
Detection Method Mass Spectrometry
Workflow Step Calibration
Quantity 24 tubes