Learn More
Thermo Scientific™ Pierce™ DMA Crosslinker
Product Code. 10752725 Shop All Thermo Scientific Products
Click to view available options
Quantity:
1 g
Unit Size:
1g
Product Code. 10752725
Brand: Thermo Scientific™ 20660
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Crosslink amines to amines with dimethyl adipimidate (CAS 14620-72-5); composed of imidoester groups and a 6-atom, membrane-permeable spacer arm.
Thermo Scientific™ Pierce DMA is dimethyl adipimidate, a membrane-permeable crosslinker that contains an amine-reactive imidoester group at each end of a 6-atom spacer arm.
Highlights:
  • Water-soluble
  • Reactive groups: imidoester (homobifunctional)
  • Reactive toward: amino groups
TRUSTED_SUSTAINABILITY
Specifications
Chemical Reactivity Amine-to-Amine
Cleavable No
Description Pierce DMA
Formulation Dimethyl adipimidate-2HCl; powder-granule solid
Membrane Permeable Yes
Molecular Weight (g/mol) 245.15
PEGylated No
Sufficient For 4079mL of typical 1X (1mM) reaction solution, or approx. 500 uses at 2mg reagent per use
Spacer Arm Length 8.6
Quantity 1 g
Solubility Water