Learn More
Thermo Scientific™ Pierce™Dimethylsulfoxide (DMSO), Sequencing Grade
Product Code. 10156644 Shop All Thermo Scientific Products
Click to view available options
Quantity:
950 mL
Unit Size:
Each
Product Code. 10156644
Brand: Thermo Scientific™ TS20688
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
A sequencing-grade DMSO preparation with high purity and consistent absorptivity properties for use in HPLC and mass spectrometry methods.
Thermo Scientific™ Pierce™ Dimethylsulfoxide (DMSO) is a sequencing-grade preparation with high purity for use in traditional amino acid analyzers and other high-volume applications.
This sequencing-grade dimethylsulfoxide is >99.5% pure DMSO which is an excellent choice for protein and peptide applications that require the water-miscible solvent. The purity test ensures that preparation is essentially devoid of amines. This DMSO is packaged in solvent-rinsed amber glass bottles and sealed with PTFE-lined fluorocarbon caps for ultimate protection.
TRUSTED_SUSTAINABILITY
Specifications
Hazards Flammable
Item Name Dimethylsulfoxide (DMSO)
Storage Room Temperature
Form Liquid
Quantity 950 mL
Formulation 99.5% Dimethylsulfoxide
Product Line Pierce
For Use With LC/MS
compliance-icons compliance-icons compliance-icons
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H361-Suspected of damaging fertility or the unborn child.
  • H373-May cause damage to organs.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
Supplemental information
  • MIXTURE LIST-Contains : DMSO