Learn More
Thermo Scientific™ GalNAz (N-azidoacetylgalactosamine tetraacylated)
Shop All Thermo Scientific ProductsDescription
Thermo Scientific™ Pierce Azido-Sugars (GlcNAz, GalNAz and ManNAz) are azide-labeled sugars that provide a highly specific approach for studying glycoproteins through in vivo metabolic labeling and chemoselective ligation.
Highlights:
Bioorthogonal - the azido group is small, nonreactive and absent from living systems; as such the azido-sugar compounds do not interfere with endogenous cellular pathways and substitute for their naturally occurring analogs
Compatible - reaction chemistry with phosphine compounds occurs effectively in simple buffer conditions; requires no accessory reagents such as copper or reducing agents
Chemoselective - Azide and phosphine groups do not react or interfere with components of biological samples but conjugate to one another with high efficiency
Versatile - Azide tag can be targeted for detection, immobilization, conjugation or affinity purification depending on which phosphine-activated compound it is reacted with
Specifications
Specifications
| Description | Pierce GalNAz |
| Product Type | Azide-labeled Sugar |
| Quantity | 5 mg |
| Content And Storage | Store at -20°C. Shipped at ambient temperature. |
| Product Line | Pierce |
| Chemical Reactivity | Phosphine, Alkyne |
| Reactive Moiety | Amine, Azide |
| Solubility | DMSO (Dimethylsulfoxide) |
| Label or Dye | Azide |
| Label Type | Staudinger Chemistry Label, Click Chemistry Label |