missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Picolinic acid, chromium salt, MP Biomedicals™
Shop All MP Biomedicals Products
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| CAS | 14639-25-9 |
| Color | Red |
| Molecular Formula | C18H12CrN3O6 |
| Quantity | 1 g |
| Synonym | chromium picolinate, chromium iii pyridine-2-carboxylate, chromium iii picolinate, chromium tripicolinate, picolinic acid chromium iii salt, chromium, tris picolinato, unii-s71t8b8z6p, chromium iii trispicolinate, picolinic acid, chromium salt, chromium 2-pyridinecarboxylate |
| InChI Key | CBDQOLKNTOMMTL-UHFFFAOYSA-K |
| SMILES | C1=CC=NC(=C1)C(=O)[O-].C1=CC=NC(=C1)C(=O)[O-].C1=CC=NC(=C1)C(=O)[O-].[Cr+3] |
| IUPAC Name | chromium(3+);pyridine-2-carboxylate |
| Molecular Weight (g/mol) | 418.305 |
| PubChem CID | 56840998 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction