Learn More
Picolinafen-d4, TRC
Specifications
Specifications
| Chemical Name or Material | Picolinafen-d4 |
| CAS | 2469244-77-5 |
| Molecular Formula | C19D4H8F4N2O2 |
| Quantity | 1 mg |
| Synonym | AC 900001-d4, N-(4-Fluorophenyl)-2-[(3-trifluoromethylphenyl)oxy]pyridine-6-carboxamide-d4, N-(4-Fluorophenyl)-6-[3-(trifluoromethyl)phenoxy]-2-pyridinecarboxamide-d4, Sniper-d4 |
| InChI Formula | InChI=1S/C19H12F4N2O2/c20-13-7-9-14(10-8-13)24-18(26)16-5-2-6-17(25-16)27-15-4-1-3-12(11-15)19(21,22)23/h1-11H,(H,24,26)/i7D,8D,9D,10D |
| SMILES | [2H]c1c([2H])c(NC(=O)c2cccc(Oc3cccc(c3)C(F)(F)F)n2)c([2H])c([2H])c1F |
| IUPAC Name | N-(2,3,5,6-tetradeuterio-4-fluoro-phenyl)-6-[3-(trifluoromethyl)phenoxy]pyridine-2-carboxamide |
| Molecular Weight (g/mol) | 380.33 |
| Formula Weight | 380.109 |