Learn More
Picaridin, TRC
Product Code. 30012526
Change view
Click to view available options
Quantity:
10 mg
1 g
5 g
Unit Size:
10mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30012526 5 g 5g
17904695 10 mg 10mg
17971255 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30012526 Supplier Toronto Research Chemicals Supplier No. TRCP4360005G
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 119515-38-7
Molecular Formula C12 H23 N O3
Molecular Weight (g/mol) 229.32
Synonym 2-(2-Hydroxyethyl)-1-piperidinecarboxylic Acid 1-Methylpropyl Ester, 1-Methylpropyl 2-(2-hydroxyethyl)-1-piperidinecarboxylate, Autan, Autan Repel, Bayrepel, Icaridin, Isobutyl 2-(2-hydroxyethyl)-1-piperidinecarboxylate, KBR 3023, Natrapel, Picaridin, Pikaridin, Propidine
IUPAC Name butan-2-yl 2-(2-hydroxyethyl)piperidine-1-carboxylate
SMILES CCC(C)OC(=O)N1CCCCC1CCO
Specifications
Quantity 5 g
InChI Formula InChI=1S/C12H23NO3/c1-3-10(2)16-12(15)13-8-5-4-6-11(13)7-9-14/h10-11,14H,3-9H2,1-2H3
Formula Weight 229.1678
Percent Purity >95%
Chemical Name or Material Picaridin
Grade HPLC