missing translation for 'onlineSavingsMsg'
Learn More

Phthalic Anhydride, Sigma-Aldrich™

Product Code. 15888330
Change view
Click to view available options
Quantity:
100 g
1 kg
5 kg
50 kg
Unit Size:
100g
1kg
50kg
5kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15888330 50 kg 50kg
15858330 100 g 100g
15868330 1 kg 1kg
15878330 5 kg 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15888330 Supplier Sigma Aldrich Supplier No. 8005929050
 Restricted Item
Estimated Shipment: 29-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For synthesis

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 85-44-9

Specifications

Melting Point 131°C
Density 1.527 g/cm3 at 20°C
Boiling Point 285°C/1013 hPa
MDL Number MFCD00005918
Quantity 50 kg
Synonym Phthalic anhydride, Phthalic anhydride
Molecular Weight (g/mol) 148.12
Grade Research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.