missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phthalic Acid, Sigma-Aldrich™
Specifications
Specifications
| Melting Point | 191°C (closed capillary tube) |
| Boiling Point | 289°C/1013 hPa (decomposition) |
| Density | 1.59 g/cm3 at 20°C |
| Quantity | 1 kg |
| Molecular Formula | C6H4(COOH)2 |
| MDL Number | MFCD00002467 |
| Synonym | Phthalic acid, Benzene-1,2-dicarboxylic acid, Phthalic acid |
| Molecular Weight (g/mol) | 166.13 |
| Grade | Ungraded |
| Chemical Name or Material | Phthalic Acid |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?