missing translation for 'onlineSavingsMsg'
Learn More

Phthalic acid, bis-isobutyl ester 10 μg/mL in Cyclohexane, Dr. Ehrenstorfer

Product Code. 30327864
Change view
Click to view available options
Quantity:
10 mL
Unit Size:
10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30327864 10 mL 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30327864 Supplier Dr. Ehrenstorfer Supplier No. DREL16173500CY
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Phthalic acid, bis-iso-butyl ester
CAS 84-69-5
Molecular Formula C16 H22 O4
Quantity 10 mL
Synonym 1,2-Benzenedicarboxylic acid, 1,2-bis(2-methylpropyl) ester, 1,2-Benzenedicarboxylic acid, bis(2-methylpropyl) ester (9CI), Phthalic acid, diisobutyl ester (6CI,7CI,8CI), 1,2-Benzenedicarboxylic acid diisobutyl ester, 2-Methylpropyl phthalate, Bis(2-methylpropyl) o-phthalate, Bis(2-methylpropyl) phthalate, Di(2-methylpropyl) phthalate, Di(isobutyl) 1,2-benzenedicarboxylate, Di-iso-Butyl phthalate, Diisobutyl phthalate, Hexaplas M/1B, Isobutyl phthalate, NSC 15316, Palatinol IC, Reomol DiBP, Phthalic acid, bis-isobutyl ester
InChI Formula InChI=1S/C16H22O4/c1-11(2)9-19-15(17)13-7-5-6-8-14(13)16(18)20-10-12(3)4/h5-8,11-12H,9-10H2,1-4H3
SMILES CC(C)COC(=O)c1ccccc1C(=O)OCC(C)C
IUPAC Name bis(2-methylpropyl) benzene-1,2-dicarboxylate
Molecular Weight (g/mol) 278.34
Formula Weight 278.1518 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.