missing translation for 'onlineSavingsMsg'
Learn More

Phosphorus Tribromide, Sigma-Aldrich™

Product Code. 15841791
Change view
Click to view available options
Quantity:
50 mL
250 mL
Unit Size:
250mL
50mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15841791 250 mL 250mL
15831791 50 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15841791 Supplier Sigma Aldrich Supplier No. 8223210250
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For synthesis

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-60-8

Specifications

Boiling Point 173°C to 175°C/1013 hPa
Melting Point -40°C
Quantity 250 mL
Density 2.85 g/cm3 at 20°C
MDL Number MFCD00011436
Synonym Phosphorus tribromide, Phosphorus(III) bromide, Phosphorus tribromide
Molecular Weight (g/mol) 270.69
Grade Research
Chemical Name or Material Phosphorus Tribromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.