Learn More
Phosphoric Acid Tris(4-methylphenyl)-d21 Ester, TRC
Specifications
Specifications
| Chemical Name or Material | Phosphoric Acid Tris(4-methylphenyl)-d21 Ester |
| CAS | 2469244-63-9 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C21D21O4P |
| Quantity | 2.5 mg |
| Synonym | Phosphoric Acid Tri-p-tolyl-d21 Ester, NSC 2181-d21, Tri-p-cresyl Phosphate-d21, Tri-p-tolyl Phosphate-d21, Tris(4-methylphenyl) Phosphate-d21, Tris(p-cresyl) Phosphate-d21, Tris(p-methylphenyl) Phosphate-d21, p-Tolyl Phosphate-d21 |
| InChI Formula | InChI=1S/C21H21O4P/c1-16-4-10-19(11-5-16)23-26(22,24-20-12-6-17(2)7-13-20)25-21-14-8-18(3)9-15-21/h4-15H,1-3H3/i1D3,2D3,3D3,4D,5D,6D,7D,8D,9D,10D,11D,12D,13D,14D,15D |
| SMILES | O=[P](OC1=C([2H])C([2H])=C(C([2H])([2H])[2H])C([2H])=C1[2H])(OC2=C([2H])C([2H])=C(C([2H])([2H])[2H])C([2H])=C2[2H])OC3=C([2H])C([2H])=C(C([2H])([2H])[2H])C([2H])=C3[2H] |
| IUPAC Name | tris(4-(methyl-d3)phenyl-2,3,5,6-d4) phosphate |
| Molecular Weight (g/mol) | 389.49 |
| Show More |