missing translation for 'onlineSavingsMsg'
Learn More

Phosphocholine Chloride Calcium Salt Tetrahydrate, TRC

Product Code. 30135290
Change view
Click to view available options
Quantity:
10 g
25 g
Unit Size:
10g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135290 25 g 25g
30011244 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30135290 Supplier Toronto Research Chemicals Supplier No. TRCP35450025G
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 72556-74-2
Molecular Formula C5 H13 N O4 P . Ca . Cl . 4 H2 O
Molecular Weight (g/mol) 329.73
Synonym Ethanaminium, N,N,N-trimethyl-2-(phosphonooxy)-, chloride, calcium salt, hydrate (1:1:1:4) (ACI), Ethanaminium, N,N,N-trimethyl-2-(phosphonooxy)-, chloride, calcium salt (1:1), tetrahydrate (9CI), N,N,N-Trimethyl-2-(phosphonooxy)ethanaminium chloride calcium salt hydrate (1:1:1:4) (ACI), Calcium phosphorylcholine tetrahydrate, Calcium phosphorylcholine chloride tetrahydrate, Phosphorylcholine calcium salt tetrahydrate, Phosphorylcholine chloride calcium salt tetrahydrate
IUPAC Name calcium;2-(trimethylazaniumyl)ethyl phosphate;chloride;tetrahydrate
SMILES O.O.O.O.[Cl-].[Ca+2].C[N+](C)(C)CCOP(=O)([O-])[O-]

Specifications

Quantity 25 g
InChI Formula InChI=1S/C5H14NO4P.Ca.ClH.4H2O/c1-6(2,3)4-5-10-11(7,8)9;;;;;;/h4-5H2,1-3H3,(H-,7,8,9);;1H;4*1H2/q;+2;;;;;/p-2
Formula Weight 329.0319
Chemical Name or Material Phosphocholine Chloride Calcium Salt Tetrahydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.