missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Phospho-TRAF7 (Ser61) Polyclonal Antibody
GREENER_CHOICE

Product Code. 16661765
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16661765 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16661765 Supplier Invitrogen™ Supplier No. PA5105321

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of TRAF7 only when phosphorylated at Ser61.

TRAFs are signal transducers for members of TNFR superfamily. These are critically involved in activation of MAP3K family members involved in NF-kappaB and JNK activation and apoptosis. TRAF7, a novel 670 A. A protein, is a human homolog of mouse Traf7 andconsists of an N-terminal RING finger domain, central TRAF domain, a coiled-coil motif and 7 WD40 repeats at the C-terminus. It has E3-ubiquitin ligase activity for self-ubiquitination. TRAF7 specifically interacts with MEKK3 and mediates MEKK3-induced AP1 and CHOP activation and induces apoptosis through Caspase-independent pathways. It is a multifunctional protein involved in sumoylation and negative regulation of c-Myb, thereby regulating proliferation and differentiation of hematopoietic cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-TRAF7 (Ser61)
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene TRAF7
Gene Accession No. Q6Q0C0, Q922B6
Gene Alias DKFZp586I021; E3 ubiquitin-protein ligase TRAF7; MGC7807; RFWD1; RGD1559653; ring finger and WD repeat domain 1; ring finger and WD repeat domain 1 isoform 1; RING finger and WD repeat-containing protein 1; RING finger protein 119; RING-type E3 ubiquitin transferase TRAF7; RNF119; TNF receptor associated factor 7; TNF receptor-associated factor 7; TNF receptor-associated factor 7, E3 ubiquitin protein ligase; traf7
Gene Symbols TRAF7
Host Species Rabbit
Immunogen A synthesized peptide derived from human TRAF7(Accession Q6Q0C0), corresponding to amino acid residues around phosphorylated Ser61.
Purification Method sequential chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 224619, 360491, 84231
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.