missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-PDGF R beta (Y751) Rabbit anti-Human, Mouse, Clone: 1210B, R&D Systems™

Rabbit Monoclonal Antibody has been used in 1 publication
Brand: R&D Systems MAB9027
This item is not returnable.
View return policy
Description
PDGF R beta Monoclonal specifically detects PDGF R beta in Human, Mouse samples. It is validated for Western Blot, Simple Western, Immunocytochemistry.Specifications
| PDGF R beta | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with Sodium Azide | |
| beta-type platelet-derived growth factor receptor, CD140 antigen-like family member B, CD140b, CD140b antigen, EC 2.7.10, EC 2.7.10.1, JTK12, PDGFR, PDGFR1, PDGFRB, PDGF-R-beta, platelet-derived growth factor receptor, beta polypeptide | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 5159 | |
| Reconstitute at 0.5 mg/mL in sterile PBS | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG |
| Western Blot, Immunocytochemistry | |
| 1210B | |
| Western Blot 0.1 ug/mL, Simple Western 1 ug/mL, Immunocytochemistry 5-25 ug/mL | |
| P09619 | |
| PDGFRB | |
| Phosphopeptide containing the human PDGF R beta (Y751) site Accession # P09619 | |
| 100 μg | |
| Primary | |
| Detects human and mouse PDGF R beta when phosphorylated at Y751 in Western blots. | |
| Human, Mouse | |
| Purified |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?