missing translation for 'onlineSavingsMsg'
Learn More

Phospho-AKT1 (Ser473) Rabbit anti-Human, Mouse, CoraLite 647, Clone: 2M10, Proteintech

Rabbit Recombinant Antibody

145.00 EUR - 450.00 EUR

Specifications

Antigen Phospho-AKT1 (Ser473)
Clone 2M10
Applications Flow Cytometry
Conjugate CoraLite 647
Form Liquid
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17721479
View Documents
Proteintech
CL647-80462-20UL
20 μL
145.00 EUR
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
17609996
View Documents
Proteintech
CL647-80462-100UL
100 μL
450.00 EUR
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Phospho-AKT1 (Ser473) antibody apecifically detetects Phospho-AKT1 (Ser473) antigen in Human, Mouse Samples. It is validated for Flow Cytometry.
Specifications

Specifications

Phospho-AKT1 (Ser473)
Flow Cytometry
Liquid
Human, Mouse
NM_005163
AKT1
IgG
Protein A purification
2M10
CoraLite 647
Rabbit
PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3.
207
Peptide
Primary
Store at -20°C. Avoid exposure to light.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.