missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-AKT1 (Ser473) Rabbit anti-Human, Mouse, CoraLite 647, Clone: 2M10, Proteintech
Rabbit Recombinant Antibody
Brand: Proteintech CL647-80462-20UL
This item is not returnable.
View return policy
Description
Phospho-AKT1 (Ser473) antibody apecifically detetects Phospho-AKT1 (Ser473) antigen in Human, Mouse Samples. It is validated for Flow Cytometry.Specifications
| Phospho-AKT1 (Ser473) | |
| 2M10 | |
| CoraLite 647 | |
| NM_005163 | |
| Rabbit | |
| Protein A purification | |
| Primary | |
| Human, Mouse | |
| Liquid |
| Flow Cytometry | |
| 1000 ug/mL | |
| PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3. | |
| AKT1 | |
| Peptide | |
| 20 μL | |
| 207 | |
| Store at -20°C. Avoid exposure to light. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction