Learn More
Medicago Phosphate Buffered Saline Tablets, pH 7.4
Product Code. 11340439
Change view
Click to view available options
Quantity:
100 x 1000 mL tablets
100 x 500 mL tablets
Unit Size:
Pack of 100
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11340439 100 x 500 mL tablets Pack of 100
11330439 100 x 1000 mL tablets Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11340439 Supplier Medicago Supplier No. 098912100
  • ONLINE EXCLUSIVE PRICE
    153.00 EUR
    136.57 EUR / Pack of 100
    Save 16.43 EUR
    10% Off
 Restricted Item
Estimated Shipment: 28-10-2026
Specifically developed for immunological and microbiological laboratories. Medicago Phosphate Buffered Saline Tablets, pH 7.4 are provided as pre-weighed tablets with a final volume of 500 or 1000ml per tablet.
Among biological buffers, PBS is one of the most commonly used. The buffer is isotonic and non-toxic to cells and has the ability to maintain their osmolarity. The buffer is suitable for washing procedures in cell cultures and for immunoassays such as ELISA and immuno-histochemical procedures. It is often used for sample dilution in molecular biology and as protein diluent in Western blotting. Furthermore, the buffer can function as an equilibrator for gel filter columns.
  • Formulated from analytical grade reagents
  • Autoclavable
  • Avaliable as convenient tablets that are ready to use in minutes
Specifications
Color White
For Use With (Application) Immunoassays, tissue and cell culture procedures, immuno-histochemical procedures, microbiological procedures, and sample dilution
pH 7.4
Physical Form Tablet
Quantity 100 x 500 mL tablets
Grade Analytical Grade