missing translation for 'onlineSavingsMsg'
Learn More

Phorbol 12-myristate 13-acetate, >99.5%, For HPLC analysis, MP Biomedicals™

Product Code. 11497072 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11497072 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11497072 Supplier MP Biomedicals Supplier No. 0215186405
  • ONLINE EXCLUSIVE PRICE
    407.00 EUR
    387.45 EUR / 5mg
    Save 19.55 EUR
    4% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 16561-29-8
Color Colorless
Assay Percent Range >99%
Molecular Formula C36H56O8
MDL Number MFCD00036736
Quantity 5 mg
Synonym phorbol 12-myristate 13-acetate, phorbol ester, 12-o-tetradecanoylphorbol-13-acetate, 12-o-tetradecanoylphorbol 13-acetate, factor a1, tetradecanoylphorbol acetate, 12-tetradecanoylphorbol 13-acetate, factor a1 croton oil, phorbol 13-acetate 12-myristate, phorbol-12-myristate-13-acetate
InChI Key PHEDXBVPIONUQT-RGYGYFBISA-N
SMILES CCCCCCCCCCCCCC(=O)O[C@@H]1[C@@H](C)[C@]2(O)[C@@H]3C=C(C)C(=O)[C@@]3(O)CC(CO)=C[C@H]2[C@@H]2C(C)(C)[C@]12OC(C)=O
IUPAC Name (1S,2S,6R,10S,11R,13S,14R,15R)-13-(acetyloxy)-1,6-dihydroxy-8-(hydroxymethyl)-4,12,12,15-tetramethyl-5-oxotetracyclo[8.5.0.0²,⁶.0¹¹,¹³]pentadeca-3,8-dien-14-yl tetradecanoate
Molecular Weight (g/mol) 616.84
PubChem CID 27924
ChEBI CHEBI:37537
Formula Weight 616.84
Percent Purity 0.995
Chemical Name or Material Phorbol 12-myristate 13-acetate, For HPLC analysis
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.