Learn More
uvex Pheos B Safety Helmet
Product Code. 17594106 Shop All Uvex Products
Change view
Click to view available options
Color:
White
Unit Size:
Pack of 25
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
17594106 White Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17594106 Supplier uvex Supplier No. 9772020
  • ONLINE EXCLUSIVE PRICE
    305.00 EUR
    287.99 EUR / Pack of 25
    Save 17.01 EUR
    5% Off
Estimated Shipment: 05-01-2027
Lightweight safety helmets that feature a sporty design and offer maximum wearer comfort.
  • Lightweight helmet in a sporty design
  • Three variable air vents, one ventilation zone on the forehead and two at the back, for perfect air circulation
  • Adapter systems for uvex visors, uvex earmuffs, and helmet lamps
  • Conventional 6-point textile suspension harness for a comfortable fit
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance EN 397
Color White
For Use With (Equipment) Hard Hats
Material HDPE
Model 9772
Product Type Industrial Safety Helmet
Size 52 to 61 cm
Size (Metric) 51 to 61 cm
Style Long Brim
Ventilated Ventilated
Head Style Front Brim