missing translation for 'onlineSavingsMsg'
Learn More

Phenylacetaldehyde, 99.5%, For GC analysis, MP Biomedicals™

Product Code. 11433512 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11433512 500 mL 500mL
11423512 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11433512 Supplier MP Biomedicals Supplier No. 0215614590
  • ONLINE EXCLUSIVE PRICE
    433.00 EUR
    411.60 EUR / 500mL
    Save 21.40 EUR
    4% Off
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Phenylacetaldehyde ≥95%, Clear Liquid

  • It is used for the preparation of fragrances and polymers like polyesters, which find application as a rate controlling additive in polymerization reactions.
  • It is an active component of fragrances and floral scent due to its honey-like sweet character and finds application in flavored cigarettes and beverages.
  • It is an insect attractant and utilized in blacklight trap for pests. It is also used as a building block in the synthesis of more complex chemicals, such as resmethrin.
  • Furthermore, it is used in association with acetic anhydride and allyltrimethylsilane in three-component coupling process catalyzed by LiBF4 providing homoallylic acetates.
  • A naturally occurring aromatic compound. Contains varying amounts of poly(styrene oxide).

Chemical Identifiers

CAS 122-78-1
Molecular Formula C8H8O
Molecular Weight (g/mol) 120.15
MDL Number MFCD00006993
InChI Key DTUQWGWMVIHBKE-UHFFFAOYSA-N
PubChem CID 998
ChEBI CHEBI:16424
IUPAC Name 2-phenylacetaldehyde
SMILES O=CCC1=CC=CC=C1

Specifications

Color Colorless
Assay Percent Range 0.995
Quantity 500 mL
Synonym phenylacetaldehyde, benzeneacetaldehyde, hyacinthin, alpha-tolualdehyde, phenylethanal, 2-phenylethanal, phenylacetic aldehyde, alpha-toluic aldehyde, acetaldehyde, phenyl, benzylcarboxaldehyde
Formula Weight 120.15
Physical Form Liquid
Chemical Name or Material Phenylacetaldehyde, For GC analysis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.