Learn More
Phenyl Ethenesulfonate, TRC
Product Code. 30427880
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30427880 500 mg -
30421923 100 mg 100mg
30425408 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30427880 Supplier Toronto Research Chemicals Supplier No. TRCP399375500MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1562-34-1
Molecular Formula C8H8O3S
Molecular Weight (g/mol) 184.21
Synonym Phenyl Ethylenesulfonate, Phenyl Vinylsulfonate
IUPAC Name phenyl ethenesulfonate
SMILES C=CS(=O)(=O)Oc1ccccc1
Specifications
Quantity 500 mg
InChI Formula InChI=1S/C8H8O3S/c1-2-12(9,10)11-8-6-4-3-5-7-8/h2-7H,1H2
Formula Weight 184.0194
Chemical Name or Material Phenyl Ethenesulfonate