missing translation for 'onlineSavingsMsg'
Learn More

Phentolamine mesylate, MedChemExpress

Product Code. p-200099017
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16468256 100 mg 100mg
16432116 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16468256 Supplier Medchem Express Supplier No. HYB0362A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Phentolamine mesylate (Phentolamine methanesulfonate) is a reversible, non-selective, and orally active blocker of α1 and α2 adrenergic receptor that expands blood vessels to reduce peripheral vascular resistance. Phentolamine mesylate can be used for the research of pheochromocytoma-related hypertension, heart failure and erectile dysfunction.

Phentolamine mesylate (Phentolamine methanesulfonate) is a reversible, non-selective, and orally active blocker of α1 and α2 adrenergic receptor that expands blood vessels to reduce peripheral vascular resistance. Phentolamine mesylate can be used for the research of pheochromocytoma-related hypertension, heart failure and erectile dysfunction.

Specifications

Chemical Name or Material Phentolamine mesylate
CAS 65-28-1
Color White
Purity Grade Notes Research
Molecular Formula C18H23N3O4S
Quantity 100 mg
Synonym Phentolamine methanesulfonate
Solubility Information H2O : ≥ 50 mg/mL (132.46 mM)
SMILES CS(O)(=O)=O.CC1=CC=C(N(C2=CC(O)=CC=C2)CC3=NCCN3)C=C1
Molecular Weight (g/mol) 377.46
Formula Weight 377.46
Percent Purity 99.9%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.