missing translation for 'onlineSavingsMsg'
Learn More

Phenserine-d5, TRC

Product Code. 30426768
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30426768 10 mg 10mg
30410503 5 mg 5mg
30423625 1 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30426768 Supplier Toronto Research Chemicals Supplier No. TRCP31890410MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C20H18D5N3O2
Molecular Weight (g/mol) 342.45
Synonym (-)-Eseroline Phenylcarbamate-d5, (-)-N-Phenylcarbamoyleseroline-d5, (-)-Phenserine-d5, (3aS-cis)-1,2,3,3a,8,8a-Hexahydro-1,3a,8-trimethyl-pyrrolo[2,3-b]indol-5-ol Phenylcarbamate-d5 (Ester), (3aS,8aR)-1,2,3,3a,8,8a-Hexahydro-1,3a,8-trimethyl-pyrrolo[2,3-b]indol-5-ol-d5 (N-Phenylcarbamate)
SMILES O=C(OC1=CC([C@@](CCN2C)(C)[C@@]2([H])N3C)=C3C=C1)NC4=C([2H])C([2H])=C([2H])C([2H])=C4[2H]

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C20H23N3O2/c1-20-11-12-22(2)18(20)23(3)17-10-9-15(13-16(17)20)25-19(24)21-14-7-5-4-6-8-14/h4-10,13,18H,11-12H2,1-3H3,(H,21,24)/t18-,20+/m1/s1/i4D,5D,6D,7D,8D
Chemical Name or Material Phenserine-d5
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.