missing translation for 'onlineSavingsMsg'
Learn More

Phenoxybenzamine Hydrochloride, Mikromol™

Product Code. 30051823
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30051823 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30051823 Supplier Mikromol Supplier No. MM0778.00
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Phenoxybenzamine Hydrochloride
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Phenoxybenzamine Hydrochloride
CAS 63-92-3
Analyte or Component Names Phenoxybenzamine Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C18 H22 Cl N O . Cl H
Certifications/Compliance ISO 17025
Synonym Benzenemethanamine, N-(2-chloroethyl)-N-(1-methyl-2-phenoxyethyl)-, hydrochloride, Benzylamine, N-(2-chloroethyl)-N-(1-methyl-2-phenoxyethyl)-, hydrochloride, 2-(N-Benzyl-2-chloroethylamino)-1-phenoxypropane hydrochloride, Dibenzylin, Dibenzyline chloride, Dibenzyline hydrochloride, Dibenzyran, N-(2-Chloroethyl)-N-(1-methyl-2-phenoxyethyl)benzylamine, hydrochloride, N-2-Phenoxyisopropyl-N-benzyl-β-chloroethylamine hydrochloride, NSC 37448, Phenoxybenzamine chloride, Phenoxybenzamine hydrochloride
InChI Formula InChI=1S/C18H22ClNO.ClH/c1-16(15-21-18-10-6-3-7-11-18)20(13-12-19)14-17-8-4-2-5-9-17;/h2-11,16H,12-15H2,1H3;1H
SMILES Cl.CC(COc1ccccc1)N(CCCl)Cc2ccccc2
IUPAC Name N-benzyl-N-(2-chloroethyl)-1-phenoxypropan-2-amine;hydrochloride
Molecular Weight (g/mol) 340.29
Quantity 250 mg
Formula Weight 339.1157 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.