Learn More
Phenolphtalein Indicator in Ethanol, 1g/L, Chem-Lab
Product Code. 11957423
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11957423 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11957423 Supplier Chem-Lab Supplier No. CL07.0606.0100
  • ONLINE EXCLUSIVE PRICE
    64.40 EUR
    47.50 EUR / 100mL
    Save 16.90 EUR
    26% Off
 Restricted Item
Estimated Shipment: 29-10-2026
Turns red-violent in the presence of bases (at pH 8.2–10). Phenolphtalein Indicator in Ethanol, 1g/L, Chem-Lab contains 1g C20H14O4 per 1L C2H5OH. It is used as an indicator in acid-base titrations. Manufactured according to Eur. Ph. 4.4.1.1 standards.
Phenolphthalein is usually dissolved in alcohol for use in experiments. It is a weak acid, which can lose H+ ions in solution. The phenolphthalein molecule is colorless, while the phenolphthalein ion is pink. In acidic and neutral solutions, the solution is colorless. When a base is added to the phenolphthalein, more ionization takes place, causing the solution to turn a red-violet color. This solution turns this pinkish color at pH 8.2–10.0.
Specifications
CAS 77-09-8, 64-17-5
Concentration 1g C20H14O4 per 1L C2H5OH 80%
Molecular Formula C20H14O4
MDL Number MFCD00005913
Solubility Information Slightly soluble in water
InChI Key KJFMBFZCATUALV-UHFFFAOYSA-N
SMILES OC1=CC=C(C=C1)C1(OC(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1
IUPAC Name 3,3-bis(4-hydroxyphenyl)-2-benzofuran-1-one
Molecular Weight (g/mol) 318.33
PubChem CID 4764
ChEBI CHEBI:34914
Formula Weight 46.07g/mol
Grade Eu. Ph. Chapter 4(4.1.1)
Odor Alcoholic
Density 0.86g/mL
pH Color Changes Colorless to red-violet at pH 8.2–10.0
Solvent or Matrix Ethanol 80%
pH ±7
Quantity 100 mL
Physical Form Liquid
Chemical Name or Material Phenolphtalein
Show More Show Less
compliance-icons
Product Identifier
  • Phenolphtalein indicator in Ethanol
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
Supplemental information
  • MIXTURE LIST-Contain: ethanol