missing translation for 'onlineSavingsMsg'
Learn More

Phenolphtalein Indicator in Ethanol, 10g/L, Chem-Lab

Product Code. 11952505
Change view
Click to view available options
Quantity:
250 mL
Unit Size:
250mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11952505 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11952505 Supplier Chem-Lab Supplier No. CL0606040250
  • ONLINE EXCLUSIVE PRICE
    14.45 EUR
    11.07 EUR / 250mL
    Save 3.38 EUR
    23% Off
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Turns red-violent in the presence of bases (at pH 8.2–9.8). Phenolphtalein Indicator in Ethanol, 10g/L, Chem-Lab contains 10g C20H14O4 per 1L C2H5OH. It is used as an indicator in acid-base titrations.

Phenolphthalein is usually dissolved in alcohol for use in experiments. It is a weak acid, which can lose H+ ions in solution. The phenolphthalein molecule is colorless, while the phenolphthalein ion is pink. In acidic and neutral solutions, the solution is colorless. When a base is added to the phenolphthalein, more ionization takes place, causing the solution to turn a red-violet color. This solution turns this pinkish color at pH 8.2–9.8.

Specifications

Melting Point -114°C
CAS 77-09-8, 64-17-5
Concentration 10g C20H14O4 per 1L C2H5OH
Molecular Formula C20H14O4
MDL Number MFCD00005913
Solubility Information Slightly soluble in water
InChI Key KJFMBFZCATUALV-UHFFFAOYSA-N
SMILES OC1=CC=C(C=C1)C1(OC(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1
IUPAC Name 3,3-bis(4-hydroxyphenyl)-2-benzofuran-1-one
Molecular Weight (g/mol) 318.33
PubChem CID 4764
ChEBI CHEBI:34914
Formula Weight 46.07g/mol
Odor Alcoholic
Density 0.79g/mL
pH Color Changes Colorless to red-violet at pH 8.2–9.8
Solvent or Matrix Ethanol
Quantity 250 mL
Physical Form Liquid
Chemical Name or Material Phenolphtalein
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Phenolphtalein 10 g/l in ethanol indicator solution
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 1B
  • Flammable liquid Category 2
  • Germ cell mutagenicity Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H341-Suspected of causing genetic defects.
  • H350-May cause cancer.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: phenolphtalein indicator, ethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.