Learn More
Phenol Red, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
Product Code. 15548824 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Wide Mouth Amber Glass Bottle
Unit Size:
25mL
5g
Product Code. Quantity Packaging unitSize
15548824 25 g Wide Mouth Amber Glass Bottle 25mL
15538824 5 g Wide Mouth Amber Glass Bottle 5g
2 options
Product Code. 15548824
Brand: Avantor J.T.Baker T25403
High quality chemicals for laboratory and specialized industrial use
Chemical Identifiers
CAS 143-74-8
Molecular Formula C19H14O5S
Molecular Weight (g/mol) 354.38
MDL Number MFCD00003552
InChI Key BELBBZDIHDAJOR-UHFFFAOYSA-N
PubChem CID 4766
ChEBI CHEBI:31991
IUPAC Name 4-[3-(4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenol
SMILES OC1=CC=C(C=C1)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1
Specifications
Formula Weight 354.39
Grade BAKER ANALYZED™ ACS Reagent
Packaging Wide Mouth Amber Glass Bottle
Quantity 25 g
Chemical Name or Material Phenol Red