accessibility menu, dialog, popup

UserName

OHOSOOHO

CAS RN 143-74-8

IUPAC Name: 3,3-bis(4-hydroxyphenyl)-3H-2,1λ⁶-benzoxathiole-1,1-dione
Synonyms: phenol red phenolsulphonphthalein sulphental phenolsulfonphthalein preparation phenolsulfonphthalein (substance) 酚磺酞 sulphonthal PSP sulfonphthal phenolsulfonphthalein
Molecular Weight (g/mol): 354.38
Molecular Formula: C19H14O5S
InChi Key: BELBBZDIHDAJOR-UHFFFAOYSA-N
SMILES: OC1=CC=C(C=C1)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1
Quantity 
  • (1)
  • (3)
  • (7)
  • (8)
  • (1)
  • (8)
  • (1)
  • (1)
Grade 
  • (3)
  • (8)
  • (2)
  • (3)
  • (5)
Color 
  • (3)
  • (3)
  • (3)
  • (3)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (2)
  • (2)
  • (5)
  • (6)
  • (6)
  • (3)
  • (5)
  • (1)
  • (3)
  • (7)
  • (8)
  • (1)
  • (8)
  • (1)
  • (1)
  • (3)
  • (8)
  • (2)
  • (3)
  • (5)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)