Learn More
Phenmetetrazine Hydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | Phenmetetrazine Hydrochloride |
| Molecular Formula | C13 H19 N O . H Cl |
| Quantity | 25 mg |
| Synonym | 4-Ethyl-3-methyl-2-phenyl-morpholine Hydrochloride |
| InChI Formula | InChI=1S/C13H19NO.ClH/c1-3-14-9-10-15-13(11(14)2)12-7-5-4-6-8-12;/h4-8,11,13H,3,9-10H2,1-2H3;1H |
| SMILES | Cl.CCN1CCOC(C1C)c2ccccc2 |
| IUPAC Name | 4-ethyl-3-methyl-2-phenylmorpholine;hydrochloride |
| Molecular Weight (g/mol) | 241.757 |
| Formula Weight | 241.123 |