missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenazopyridine Hydrochloride, Mikromol™
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Description
Phenazopyridine Hydrochloride
Specifications
Specifications
| Chemical Name or Material | Phenazopyridine Hydrochloride |
| CAS | 136-40-3 |
| Analyte or Component Names | Phenazopyridine Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C11 H11 N5 . Cl H |
| Certifications/Compliance | ISO 17025 |
| Synonym | 2,6-Pyridinediamine, 3-(2-phenyldiazenyl)-, hydrochloride (1:1), 2,6-Pyridinediamine, 3-(phenylazo)-, monohydrochloride (9CI), Pyridine, 2,6-diamino-3-(phenylazo)-, monohydrochloride (8CI), 2,6-Diamino-3-phenylazopyridine hydrochloride, 2,6-Diamino-3-phenylazopyridine monohydrochloride, Azodyne, Bisteril, Diridone, Mallophene, NC 150, Phenazodine, Phenazopyridine hydrochloride, Phenazopyridinium chloride, Phenylazo tablets, Pirid, Pyridacil, Pyridiate, Pyridium, Pyripyridium, Sedural, Uridinal, Urisept, Urodine, W 1655, beta-Phenylazo-alpha,alpha'-diaminopyridine hydrochloride |
| InChI Formula | InChI=1S/C11H11N5.ClH/c12-10-7-6-9(11(13)14-10)16-15-8-4-2-1-3-5-8;/h1-7H,(H4,12,13,14);1H/b16-15+; |
| SMILES | Cl.Nc1ccc(N=Nc2ccccc2)c(N)n1 |
| IUPAC Name | 3-[(E)-phenyldiazenyl]pyridine-2,6-diamine;hydrochloride |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction