missing translation for 'onlineSavingsMsg'
Learn More

Phenazine Ethosulfate, MP Biomedicals

Product Code. 11495833 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11495833 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11495833 Supplier MP Biomedicals Supplier No. 02100956.5
  • ONLINE EXCLUSIVE PRICE
    68.35 EUR
    65.10 EUR / 500mg
    Save 3.25 EUR
    4% Off
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

  • Phenazine ethosulfate is an intermediate
  • Used to detect nitric oxide reducatase activity in concert with ascorbic acid

Specifications

CAS 10510-77-7
Melting Point 173°C to 175°C (literature)
Color Yellow
Molecular Formula C16H18N2O4S
Linear Formula C16H18N2O4S
MDL Number MFCD00050286
Quantity 500 mg
Synonym phenazine ethosulfate, 5-ethylphenazine, n-ethylphenazonium ethosulfate, 5-ethylphenazinium ethyl sulphate, 5-ethylphenazin-5-ium ethyl sulfate, 5-ethylphenazinium ethylsulfate, phenazinium, 5-ethyl-, ethyl sulfate, 5-ethylphenazine, ethyl sulfate salt, ccris 5619, n-ethyldibenzopyrazine ethyl sulfate salt
Solubility Information Soluble in water (10mg/mL - clear, yellow solution) or ethanol (50mg/mL - clear, dark yellow to orange solution)
InChI Key VDJKJPMLWJWQIH-UHFFFAOYSA-M
SMILES CC[N+]1=C2C=CC=CC2=NC3=CC=CC=C31.CCOS(=O)(=O)[O-]
IUPAC Name 5-ethylphenazin-5-ium;ethyl sulfate
Molecular Weight (g/mol) 334.39
PubChem CID 82688
Formula Weight 334.4
Percent Purity ∼95%
Physical Form Powder
Chemical Name or Material PHENAZINE ETHOSULFATE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.