Learn More
Phenaglycodol, TRC
Product Code. 30426174
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30426174 5 mg 5mg
30411333 50 mg 50mg
30424330 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30426174 Supplier Toronto Research Chemicals Supplier No. TRCP2947255MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 79-93-6
Molecular Formula C11H15ClO2
Molecular Weight (g/mol) 214.69
Synonym 2-(4-Chlorophenyl)-3-methyl-2,3-butanediol, 2-p-Chlorophenyl-3-methyl-2,3-butanediol, Acalmid, Acalo, Alterton, Felixyn, NSC 169451, NSC 63894, Sinforil, Stesil, Ultran
IUPAC Name 2-(4-chlorophenyl)-3-methylbutane-2,3-diol
SMILES CC(C)(O)C(C)(O)c1ccc(Cl)cc1
Specifications
Quantity 5 mg
InChI Formula InChI=1S/C11H15ClO2/c1-10(2,13)11(3,14)8-4-6-9(12)7-5-8/h4-7,13-14H,1-3H3
Formula Weight 214.0761
Chemical Name or Material Phenaglycodol