Learn More
Phellodendrine Chloride, TRC
Product Code. 30424689
Change view
Click to view available options
Quantity:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30424689 10 mg
30422975 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30424689 Supplier Toronto Research Chemicals Supplier No. TRCP29456510MG
 Restricted Item
Estimated Shipment: 12-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 104112-82-5
Molecular Formula C20H24ClNO4
Molecular Weight (g/mol) 377.87
Synonym (7S-cis)5,8,13,13a-Tetrahydro-2,11-dihydroxy-3,10-dimethoxy-7-methyl-6H-Dibenzo[a,g]quinolizinium Chloride,
IUPAC Name (7S,13aS)-3,10-dimethoxy-7-methyl-6,8,13,13a-tetrahydro-5H-isoquinolino[3,2-a]isoquinolin-7-ium-2,11-diol;chloride
SMILES [Cl-].COc1cc2CC[N@@+]3(C)Cc4cc(OC)c(O)cc4C[C@H]3c2cc1O
Specifications
Quantity 10 mg
InChI Formula InChI=1S/C20H23NO4.ClH/c1-21-5-4-12-8-19(24-2)18(23)10-15(12)16(21)6-13-7-17(22)20(25-3)9-14(13)11-21;/h7-10,16H,4-6,11H2,1-3H3,(H-,22,23);1H/t16-,21-;/m0./s1
Formula Weight 377.1394
Chemical Name or Material Phellodendrine Chloride