missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phe-Arg-β-naphthylamide dihydrochloride, >99%, For TLC analysis, 85.4%, MP Biomedicals™
Shop All MP Biomedicals Products
Click to view available options
Quantity:
10mg
25mg
50mg
250mg
Unit Size:
10mg
250mg
25mg
50mg
Specifications
Specifications
| Purity | >99% |
| Assay | 0.854 |
| CAS | 100929-99-5 |
| Color | White |
| Content And Storage | Store at 0°C. |
| Format | Powder |
| Molecular Weight (g/mol) | 519.471 |
| Product Type | Phe-Arg-β-naphthylamide dihydrochloride |
| Quantity | 250mg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction