missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PHD1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
149.35 EUR - 422.30 EUR
Specifications
| Antigen | PHD1 |
|---|---|
| Concentration | 0.33 mg/mL |
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16890974
|
Proteintech
12984-1-AP-20UL |
20 μL |
149.35 EUR
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16880974
|
Proteintech
12984-1-AP-150UL |
150 μL |
422.30 EUR
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The hypoxia inducible factor (HIF) is a transcriptional complex which is involved in oxygen homeostasis. At normal oxygen levels, the alpha subunit of HIF is targeted for degration by prolyl hydroxylation. This gene encodes an enzyme responsible for this posttranslational modification. Multiple alternatively spliced variants, encoding the same protein, have been identified.Specifications
| PHD1 | |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Rat, Mouse, Human | |
| Q6AYU4, Q91YE2, Q96KS0 | |
| 112398, 112406, 308457 | |
| PHD1 Fusion Protein Ag3616 | |
| Primary | |
| -20°C | |
| EGLN2 |
| 0.33 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Egl nine homolog 2, EGLN2, EIT6, Estrogen induced tag 6, HIF PH1, HIF prolyl hydroxylase 1, HIFPH1, HPH 1, HPH 3, PHD1 | |
| Egln2 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title