missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PHD1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12984-1-AP-20UL
This item is not returnable.
View return policy
Description
The hypoxia inducible factor (HIF) is a transcriptional complex which is involved in oxygen homeostasis. At normal oxygen levels, the alpha subunit of HIF is targeted for degration by prolyl hydroxylation. This gene encodes an enzyme responsible for this posttranslational modification. Multiple alternatively spliced variants, encoding the same protein, have been identified.Specifications
| PHD1 | |
| Polyclonal | |
| Unconjugated | |
| EGLN2 | |
| Egl nine homolog 2, EGLN2, EIT6, Estrogen induced tag 6, HIF PH1, HIF prolyl hydroxylase 1, HIFPH1, HPH 1, HPH 3, PHD1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 112398, 112406, 308457 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q6AYU4, Q91YE2, Q96KS0 | |
| Egln2 | |
| PHD1 Fusion Protein Ag3616 | |
| 20 μL | |
| Primary | |
| Rat, Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction