Learn More
Carl Zeiss™ Phase Telescope
Product Code. 11815335
Change view
Click to view available options
For Use With (Equipment):
Axiovert
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
11815335 Axiovert Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11815335 Supplier Carl Zeiss™ Supplier No. 4448309902000
  • ONLINE EXCLUSIVE PRICE
    318.15 EUR
    244.00 EUR / Pack of 1
    Save 74.15 EUR
    23% Off
Estimated Shipment: 23-11-2026
An innovative inverted microscope with superior optical performance across a wide range of functions
An innovative inverted microscope with superior optical performance across a wide range of functions.
  • An inverted microscope for cell and molecular biology
  • Ideal for examining living cells
  • Easy to operate, robust, ergonomic design and features quality optics
  • Efficient illumination: halogen lamp with more than an 800hr life
  • Two effective techniques for thin specimens: Brightfield and phase contrast
  • Two effective techniques for thick specimens: VAREL (variable relief contrast) and PlasDIC (differential interference contrast)
  • Exceptionally high fluorescence standards using the Axiovert 40 CFL
  • Flexible upgrading for future demands
Specifications
Type Phase Telescope
For Use With (Equipment) Axiovert
Quantity 1 Ea.