missing translation for 'onlineSavingsMsg'
Learn More

PharmaVetResiMix 7 Nitroimidazoles-v13 10 μg/mL in Methanol, Dr. Ehrenstorfer

Product Code. 30326366
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30326366 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30326366 Supplier Dr. Ehrenstorfer Supplier No. DREA30000037ME
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Carnidazole∣Dimetridazole∣Dimetridazole-2-hydroxy∣Furazolidone∣Ipronidazole∣Ipronidazole-hydroxy∣Metronidazole∣Metronidazole-hydroxy∣Nitrofurazone∣Ornidazole∣Ronidazole (RNZ)∣Ternidazole Hydrochloride∣Tinidazole
CAS 42116-76-7, 551-92-8, 936-05-0, 67-45-8, 14885-29-1, 35175-14-5
Analyte or Component Names Carnidazole∣Dimetridazole∣Dimetridazole-2-hydroxy∣Furazolidone∣Ipronidazole∣Ipronidazole-hydroxy∣Metronidazole∣Metronidazole-hydroxy∣Nitrofurazone∣Ornidazole∣Ronidazole (RNZ)∣Ternidazole Hydrochloride∣Tinidazole
Shipping Condition +20°C
Certifications/Compliance ISO/IEC 17025
Solvent or Matrix Methanol
Quantity 1 mL
Concentration or Composition (by Analyte or Components) Carnidazole 10 μg/mL∣Dimetridazole 10 μg/mL∣Dimetridazole-2-hydroxy 10 μg/mL∣Ipronidazole 10 μg/mL∣Ipronidazole-hydroxy 10 μg/mL∣Metronidazole 10 μg/mL∣Nitrofurazone 10 μg/mL∣Ornidazole 10 μg/mL∣Ronidazole (RNZ) 10 μg/mL∣Ternidazole Hydrochloride 10 μg/mL∣Tinidazole 10 μg/mL∣Furazolidone 10.1 μg/mL∣Metronidazole-hydroxy 10.1 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.