Learn More
Pharacine, TRC
Product Code. 17949064
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17949064 50 mg 50mg
17947334 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17949064 Supplier Toronto Research Chemicals Supplier No. TRCP29455050MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 63440-93-7
Molecular Formula C24H24O8
Molecular Weight (g/mol) 440.44
Synonym 3,8,15,20-Tetraoxatricyclo[20.2.2.210,13]octacosa-10,12,22,24,25,27-hexaene-2,9,14,21-tetrone, 1,4-Butanediol-terephthaloyl chloride cyclic dimer, Pharacin, Pharacine, X-Guard VCA 1, Polybutylene Terephthalate Cyclic Dimer, PBT Cyclic Dimer
IUPAC Name 3,8,15,20-tetraoxatricyclo[20.2.2.210,13]octacosa-1(25),10,12,22(26),23,27-hexaene-2,9,14,21-tetrone
SMILES O=C1OCCCCOC(=O)c2ccc(cc2)C(=O)OCCCCOC(=O)c3ccc1cc3
Specifications
Quantity 50 mg
InChI Formula InChI=1S/C24H24O8/c25-21-17-5-7-19(8-6-17)23(27)31-15-3-4-16-32-24(28)20-11-9-18(10-12-20)22(26)30-14-2-1-13-29-21/h5-12H,1-4,13-16H2
Formula Weight 440.1471
Percent Purity >95%
Chemical Name or Material Pharacine
Grade HPLC